Learn More
Aviva Systems Biology Corp ZNF192 Antibody - N-terminal region (ARP37750_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37750_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF192. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF192. Alternate names: LD5-1, ZKSCAN8, ZNF192ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Pig
Catalog No. 50-014-56419