Learn More
Aviva Systems Biology Corp ZNF192 Antibody - middle region (ARP38775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF192. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF192. Alternate names: LD5-1, ZKSCAN8, ZNF192ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56793