Learn More
Aviva Systems Biology Corp ZNF193 Antibody - middle region (ARP35793_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35793_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF193. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF193. Alternate names: PRD51, ZSCAN9, ZNF193ApplicationWB. Species reactivity: Cow, Human, Pig
Catalog No. 50-014-55856