Learn More
Aviva Systems Biology Corp ZNF195 Antibody - N-terminal region (ARP35834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF195. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF195. Alternate names: DKFZp666D035, HRF1, ZNFP104ApplicationWB. Species reactivity: Human
Catalog No. 50-014-55884