Learn More
Aviva Systems Biology Corp ZNF195 Antibody - N-terminal region (ARP38915_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38915_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF195. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF195. Alternate names: HRF1, ZNFP104ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-56847