Learn More
Aviva Systems Biology Corp ZNF197 Antibody - N-terminal region (ARP38893_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38893_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF197. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF197. Alternate names: P18, VHLaK, ZNF20, ZNF166, ZKSCAN9, D3S1363EApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-56838