Learn More
Aviva Systems Biology Corp ZNF197 Antibody - middle region (ARP38894_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38894_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF197. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF197. Alternate names: D3S1363E, P18, VHLaK, ZKSCAN9, ZNF166, ZNF20ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-53119