Learn More
Aviva Systems Biology Corp ZNF205 Antibody - middle region (ARP38354_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38354_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF205. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF205. Alternate names: ZNF210, Zfp13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-56619