Learn More
Aviva Systems Biology Corp ZNF213 Antibody - middle region (ARP38426_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38426_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF213. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF213. Alternate names: CR53, ZKSCAN21ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56653