Learn More
Aviva Systems Biology Corp ZNF214 Antibody - N-terminal region (ARP35858_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35858_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF214. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF214. Alternate names: BAZ1, BAZ-1ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-55892