Learn More
Aviva Systems Biology Corp ZNF219 Antibody - middle region (ARP39260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF219. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF219. Alternate names: ZFP219ApplicationWB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-56961