Learn More
Aviva Systems Biology Corp ZNF22 Antibody - C-terminal region (ARP34821_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34821_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF22. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF22. Alternate names: KOX15, HKR-T1, ZNF422, Zfp422ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55575