Learn More
Aviva Systems Biology Corp ZNF224 Antibody - middle region (ARP58369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF224. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF224. Alternate names: BMZF-2, BMZF2, KOX22, ZNF255, ZNF27ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-286-468