Learn More
Aviva Systems Biology Corp ZNF112 Antibody - N-terminal region (ARP39034_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39034_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF228. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF228. Alternate names: Zfp112, ZNF228ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-56893