Learn More
Aviva Systems Biology Corp ZNF232 Antibody - N-terminal region (ARP35880_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35880_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF232. It was validated on Western Blot. . Alternate names: ZSCAN11ApplicationWB. Species reactivity: Guinea Pig, Human
Catalog No. 50-284-478