Learn More
Aviva Systems Biology Corp ZNF236 Antibody - middle region (ARP33556_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33556_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF236. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF236. Alternate names: ZNF236A, ZNF236BApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55070