Learn More
Aviva Systems Biology Corp ZNF237 Antibody - N-terminal region (ARP34441_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34441_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF237. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF237. Alternate names: MYM, ZNF237, HSPC050, ZNF198L1ApplicationWB. Species reactivity: Cow, Guinea Pig, Human
Catalog No. 50-014-52957