Learn More
Aviva Systems Biology Corp ZNF238 Antibody - N-terminal region (ARP35799_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35799_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF238. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF238. Alternate names: C2H2-171, RP58, TAZ-1, ZBTB18, ZNF238ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55860