Learn More
Aviva Systems Biology Corp ZNF239 Antibody - middle region (ARP35785_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35785_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF239. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF239. Alternate names: HOK-2, MOK2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-55849