Learn More
Aviva Systems Biology Corp ZNF239 Antibody - N-terminal region (ARP35784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF239. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF239. Alternate names: HOK-2, MOK2ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-55848