Learn More
Aviva Systems Biology Corp ZNF24 Antibody - middle region (ARP33519_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33519_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF24. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF24. Alternate names: KOX17, RSG-A, ZNF191, ZSCAN3, Zfp191ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55053