Learn More
ABclonal Technology ZNF248 Rabbit pAb
SDP
Supplier:  ABclonal Technology A149151000UL
Encompass_Preferred
Predicted to enable DNA-binding transcription repressor activity, RNA polymerase II-specific and RNA polymerase II transcription regulatory region sequence-specific DNA binding activity. Predicted to be involved in negative regulation of transcription by RNA polymerase II. Predicted to be located in nucleus.
Catalog No. 50-001-05203