Learn More
Aviva Systems Biology Corp ZNF251 Antibody - middle region (ARP36312_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36312_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF251. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF251. Alternate names: FLJ34319ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56113