Learn More
Aviva Systems Biology Corp ZNF253 Antibody - C-terminal region (ARP39478_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39478_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF253. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF253. Alternate names: BMZF1, BMZF-1, ZNF411ApplicationWB. Species reactivity: Cow, Horse, Human, Zebrafish
Catalog No. 50-014-57050