Learn More
Aviva Systems Biology Corp ZNF256 Antibody - N-terminal region (ARP47598_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47598_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF256. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF256. Alternate names: BMZF-3, BMZF3ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59603