Learn More
Aviva Systems Biology Corp ZNF257 Antibody - N-terminal region (ARP57954_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57954_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF257. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF257. Alternate names: BMZF-4, BMZF4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62189