Learn More
Aviva Systems Biology Corp ZNF258 Antibody - middle region (ARP34839_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34839_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF258. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human ZNF258. Alternate names: MYM, ZNF258, Buster2, ZNF198L4ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Rabbit
Catalog No. 50-014-55583