Learn More
Aviva Systems Biology Corp ZPR1 Antibody - C-terminal region (ARP38400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF259. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF259. Alternate names: ZPR1ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-56638