Learn More
Aviva Systems Biology Corp ZNF265 Antibody - N-terminal region (ARP37747_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37747_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF265. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF265. Alternate names: ZIS, ZIS1, ZIS2, ZNF265ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56417