Learn More
ABclonal Technology ZNF273 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148471000UL
Encompass_Preferred
This gene is a member of the krueppel C2H2-type zinc-finger protein family and encodes a protein with 13 C2H2-type zinc fingers and a KRAB domain. This nuclear protein is involved in transcriptional regulation. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-04635