Learn More
Aviva Systems Biology Corp ZNF274 Antibody - N-terminal region (ARP31504_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31504_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF274. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF274. Alternate names: DKFZp686K08243, HFB101, ZF2, ZKSCAN19ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54247