Learn More
Aviva Systems Biology Corp ZNF276 Antibody - middle region (ARP36165_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36165_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF276. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF276. Alternate names: CENPZ, CENP-Z, ZFP276, ZNF477ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-56041