Learn More
Aviva Systems Biology Corp ZNF276 Antibody - middle region (ARP36164_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36164_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF276. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF276. Alternate names: MGC111410, MGC126480, MGC45417, ZFP276, ZNF477, CENPZ, CENP-ZApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56040