Learn More
Aviva Systems Biology Corp ZNF280A Antibody - C-terminal region (ARP50755_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50755_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF280A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF280A. Alternate names: 3'OY11.1, SUHW1, ZNF280, ZNF636ApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-014-60425