Learn More
Aviva Systems Biology Corp ZNF280C Antibody - N-terminal region (ARP39303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF280C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF280C. Alternate names: FLJ20095, SUHW3, ZNF633ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-56979