Learn More
Aviva Systems Biology Corp ZNF280D Antibody - middle region (ARP35936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF280D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF280D. Alternate names: MGC21637, MGC61687, SUHW4, ZNF634ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55933