Learn More
Aviva Systems Biology Corp ZNF282 Antibody - N-terminal region (ARP38360_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38360_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF282. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF282. Alternate names: HUB1ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56621