Learn More
Aviva Systems Biology Corp ZNF282 Antibody - C-terminal region (ARP38361_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38361_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF282. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF282. Alternate names: HUB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56622