Learn More
Aviva Systems Biology Corp ZNF284 Antibody - middle region (ARP50430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF284. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF284. Alternate names: DKFZp781F1775, ZNF284LApplicationWB. Species reactivity: Human
Catalog No. 50-014-60340