Learn More
Aviva Systems Biology Corp ZNF285A Antibody - middle region (ARP36170_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36170_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF285A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF285A. Alternate names: FLJ30747, ZNF285, ZNF285AApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-56046