Learn More
Aviva Systems Biology Corp ZNF286A Antibody - N-terminal region (ARP35974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF286A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF286A. Alternate names: KIAA1874, MGC149627, MGC156181, ZNF286ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55950