Learn More
Aviva Systems Biology Corp ZNF287 Antibody - N-terminal region (ARP39425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF287. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF287. Alternate names: MGC126536, MGC141923, ZKSCAN13ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rat
Catalog No. 50-014-57027