Learn More
Aviva Systems Biology Corp ZNF289 Antibody - N-terminal region (ARP34642_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34642_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF289. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF289. Alternate names: IRZ, ZFP289, ZNF289, NBLA10535ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55494