Learn More
Aviva Systems Biology Corp ZNF294 Antibody - N-terminal region (ARP43201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF294. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF294. Alternate names: C21orf10, C21orf98, FLJ11053, KIAA0714, RNF160, ZNF294ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63047