Learn More
Aviva Systems Biology Corp ZNF297B Antibody - N-terminal region (ARP31799_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31799_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF297B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF297B. Alternate names: ZNF-X, ZBTB22B, ZNF297BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54325