Learn More
Aviva Systems Biology Corp ZNF3 Antibody - N-terminal region (ARP33506_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33506_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF3. Alternate names: A8-51, FLJ20216, HF.12, KOX25, PP838, Zfp113ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55046