Learn More
Aviva Systems Biology Corp ZNF302 Antibody - N-terminal region (ARP39359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF302. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF302. Alternate names: HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-57001