Learn More
Aviva Systems Biology Corp ZNF304 Antibody - N-terminal region (ARP39427_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39427_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF304. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF304. Alternate names: MGC129703, MGC129704ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-284-804