Learn More
Aviva Systems Biology Corp ZNF307 Antibody - C-terminal region (ARP39385_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39385_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF307. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF307. Alternate names: ZNF307, ZNF427, P1P373C6, p373c6.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57012