Learn More
Aviva Systems Biology Corp ZNF31 Antibody - N-terminal region (ARP39855_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39855_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF31. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF31. Alternate names: KOX29, ZNF31, ZFP-31, ZNF360ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig
Catalog No. 50-014-57204